Learn More
Sino Biological Recombinant Anti-TNFR1/CD120a Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10872R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TNFR1 / CD120a / TNFRSF1A (rh TNFR1 / CD120a / TNFRSF1A; Catalog#10872-H08H; NP_001056.1; Met1-Thr211).
Catalog No. 50-240-9725