Learn More
Sino Biological Anti-TNFR1/CD120a Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10872MM1020
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNFR1 / TNFRSF1A / CD120a (rh TNFR1 / TNFRSF1A / CD120a; Catalog#10872-H08H; NP_001056.1; Met1-Thr211). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0479