Learn More
Sino Biological Anti-TNFR-2 / CD120b Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10417RP0220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TNFRSF1B / CD120b (rh TNFRSF1B; Catalog#10417-H08H; NP_001057.1; Met 1-Asp 257). TNFRSF1B specific IgG was purified by human TNFRSF1B affinity chromatography.
Catalog No. 50-241-2276