Learn More
Sino Biological Anti-DR6/TNFRSF21 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10175T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human DR6 (rh DR6; Catalog#10175-H08H; NP_055267.1; Met1-Leu350). DR6 specific IgG was purified by Human DR6 affinity chromatography.
Catalog No. 50-255-5443