Learn More
Sino Biological Human TNFRSF21 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 10175H02H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Leu 350) of human DR6 (NP_055267.1) precursor was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-0595