Learn More
Sino Biological Human TNFRSF25 / DR3 / TNFRSF12 Protein (Fc Tag) 1mg
Supplier:  Sino Biological 12095H02H1
Encompass_Preferred
A DNA sequence encoding the human TNFRSF25 (NP_683867.1) (Met1-Gln199) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6707