Learn More
Sino Biological Human TNFRSF4 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10481MCA7100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human OX40 (rh OX40; Catalog#10481-H08H; NP_003318.1; Met1-Ala216). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2819