Learn More
Sino Biological Anti-OX40/TNFRSF4 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10481MM0320
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human OX40/TNFRSF4/CD134 (rh OX40/TNFRSF4/CD134; Catalog#10481-H08H; NP_003318.1; Met1-Ala216). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5277