Learn More
Sino Biological Anti-OX40/TNFRSF4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10481RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TNFRSF4 (rh TNFRSF4; Catalog#10481-H08H; NP_003318.1; Met 1-Ala 216). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1414