Learn More
Sino Biological Anti-OX40/TNFRSF4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10481T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human OX40 / CD134 (rh OX40 / CD134; Catalog#10481-H08H; NP_003318.1; Met1-Ala216). OX40 / CD134 specific IgG was purified by Human OX40 / CD134 affinity chromatography.
Catalog No. 50-240-9476