Learn More
Sino Biological Recombinant Human OX40/TNFRSF4 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10481H49HB100
Encompass_Preferred
A DNA sequence encoding the human TNFRSF4 (NP_003318.1) (Met1-Ala216) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-9685