Learn More
Sino Biological Recombinant Human OX40/TNFRSF4 Protein (hFc & AVI Tag), Biotinylated
Supplier:  Sino Biological 10481H41HB20
Encompass_Preferred
A DNA sequence encoding the human TNFRSF4 (NP_003318.1)(Met1-Ala216) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-8061