Learn More
Sino Biological Recombinant Anti-CD30/TNFRSF8 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10777R002A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD30 / TNFRSF8 (rh CD30 / TNFRSF8; Catalog#10777-H08H; NP_001234.2; Met1-Lys379) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-1804