Learn More
Sino Biological Anti-CD30/TNFRSF8 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10777MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD30 / TNFRSF8 (rh CD30 / TNFRSF8; Catalog#10777-H08H; NP_001234.2; Met 1-Lys 379). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1733