Learn More
Sino Biological Recombinant Anti-CD30/TNFRSF8 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 10777R002P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD30 / TNFRSF8 (rh CD30 / TNFRSF8; Catalog#10777-H08H; NP_001234.2; Met1-Lys379) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-1913