Learn More
Sino Biological Anti-CD30/TNFRSF8 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10777MM05F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD30 / TNFRSF8 (rh CD30 / TNFRSF8; Catalog#10777-H08H; NP_001234.2; Met 1-Lys 379) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1736