Learn More
Sino Biological Anti-CD30/TNFRSF8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10777T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD30/TNFRSF8 (rh CD30/TNFRSF8; Catalog#10777-H08H; NP_001234.2; Met1-Lys379). CD30/TNFRSF8 specific IgG was purified by Human CD30/TNFRSF8 affinity chromatography.
Catalog No. 50-241-1916