Learn More
Sino Biological Anti-CD30/TNFRSF8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10777RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD30 / TNFRSF8 (rh CD30 / TNFRSF8; Catalog#10777-H08H; NP_001234.2; Met 1-Lys 379). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2117