Learn More
Sino Biological Recombinant Human CD30/TNFRSF8 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10777H08HB20
Encompass_Preferred
A DNA sequence encoding the Human TNFRSF8 (NP_001234.2) (Met1-Lys379) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5167