Learn More
Sino Biological Recombinant Human 4-1BB/CD137 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10041H02H1
Encompass_Preferred
A DNA sequence encoding the human TNFRSF9 (NP_001552.2) (Met1-Gln186) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5521