Learn More
Sino Biological Recombinant Human 4-1BB/CD137 Protein (ECD, hFc & AVI Tag), Biotinylated
Supplier:  Sino Biological 10041H41HB20
Encompass_Preferred
A DNA sequence encoding the human TNFRSF9 ( NP_001552.2) (Met1-Gln186) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5519