Learn More
Sino Biological Recombinant Human 4-1BB/CD137 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10041H27HB20
Encompass_Preferred
A DNA sequence encoding the human TNFRSF9 (NP_001552.2) (Met1-Gln186) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-2533