Learn More
Sino Biological Anti-TRAIL/TNFSF10 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10409T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TRAIL/TNFSF10 (rh TRAIL/TNFSF10; Catalog#10409-HNAE; NP_003801.1; Val114-Gly281). TRAIL/TNFSF10 specific IgG was purified by Human TRAIL/TNFSF10 affinity chromatography.
Catalog No. 50-255-5312