Learn More
Abcam Human TNFSF11 ELISA Kit (RANKL), 1 x 96 Tests
Supplier:  Abcam AB2138411X96TESTS
Encompass_Preferred
Human TNFSF11 ELISA Kit (RANKL) is a Sandwich (quantitative) ELISA kit for the measurement of Human TNFSF11 (RANKL) in Human in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1740277