Learn More
Sino Biological Human TNFSF11 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11682T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RANKL / OPGL / TNFSF11 / CD254 (rh RANKL / OPGL / TNFSF11 / CD254; Catalog#11682-HNCH; AAC51762.1;Gly64-Asp245). RANKL / OPGL / TNFSF11 / CD254 specific IgG was purified by Human RANKL / OPGL / TNFSF11 / CD254 affinity chromatography.
Catalog No. 50-237-3526