Learn More
Sino Biological Recombinant Human RANKL/TNFSF11 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 11682H01H50
Encompass_Preferred
A DNA sequence encoding the human TNFSF11 (AAC51762.1) (Gly64-Asp245) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-255-4620