Learn More
Sino Biological Human TNFSF11 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 11682H15H1100
Encompass_Preferred
A DNA sequence encoding the human TNFSF11 (AAC51762.1) (Gly64-Asp245) was expressed with the Fc region of rabbit IgG at the N-terminus.
Catalog No. 50-237-2951