Learn More
Sino Biological Recombinant Human APRIL/TNFSF13 Protein (AVI & His Tag), Avi-tag Biotinylated, MALS-verified
Catalog No. 502992871
Encompass_Preferred
A DNA sequence encoding the Human TNFSF13 (O75888-1) (Gln111-Leu250) was expressed with an N-terminal AVI tagged polyhistidine tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed. This construct has a trimer design.
Catalog No. 50-299-2871 Supplier Sino Biological Supplier No. 10610H44H1B20
A DNA sequence encoding the Human TNFSF13 (O75888-1) (Gln111-Leu250) was expressed with an N-terminal AVI tagged polyhistidine tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed. This construct has a trimer design.