Learn More
Sino Biological Anti-BAFF/BLyS/TNFSF13B Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10056MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human BAFF/BLyS (rh BAFF/BLyS; Catalog#10056-HNCH; Q9Y275-1; Ala134-Leu285). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2381