Learn More
Sino Biological Recombinant Anti-BAFF/BLyS/TNFSF13B Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10056R305100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human BAFF/TNFSF13B (rh BAFF/TNFSF13B; Catalog#10056-HNAE; Q9Y275-1; Ala134-Leu285).
Catalog No. 50-241-1980