Learn More
Sino Biological Recombinant Human BAFF/BLyS/TNFSF13B Protein (ECD, AVI & Fc Tag), Biotinylated, Trimer, MALS-verified
Supplier:  Sino Biological 10056H42HB100
Encompass_Preferred
A DNA sequence encoding the human TNFSF13B (NP_006564.1) (Ala134-Leu285) was expressed with a N-terminal AVI tag followed by a Fc region of human IgG1 tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed..
Catalog No. 50-255-5514