Learn More
Sino Biological Recombinant Human LIGHT/TNFSF14 Protein (ECD, AVI & Fc Tag), Biotinylated
Supplier:  Sino Biological 10386H42HB100
Encompass_Preferred
A DNA sequence encoding the human TNFSF14 (NP_001363816.1) (Asp74-Val240) was expressed with a N-terminal AVI tag followed by a Fc region of human IgG1 tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5341