Learn More
Sino Biological Recombinant Human TL1A/TNFSF15 Protein (Trimer, His & AVI Tag), Avi-tag Biotinylated, MALS-verified
Catalog No. 502993168
Encompass_Preferred
A DNA sequence encoding the Human TL1A / TNFSF15 (NP_005109.2) (Leu72-Leu251) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed. This construct has a trimer design.
Catalog No. 50-299-3168 Supplier Sino Biological Supplier No. 17049H56H2B20
A DNA sequence encoding the Human TL1A / TNFSF15 (NP_005109.2) (Leu72-Leu251) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed. This construct has a trimer design.