Learn More
KACTUSBIO INC Human TNFSF15 Trimer Protein (FSF-HM415) 500ug
Supplier:  KACTUSBIO INC FSFHM415500UG
Encompass
Recombinant Human TNFSF15 Trimer Protein is expressed from HEK293 with His tag and Flag tag at the N-Terminus.It contains Asp91-Leu251.
Catalog No. NC2737168