Learn More
Sino Biological Recombinant Human GITR Ligand/TNFSF18 Protein (His Tag)
Supplier:  Sino Biological 16080H07B100
Encompass_Preferred
A DNA sequence encoding the human TNFSF18 (AAD22634.1) (Glu52-Ser177) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-4052