Learn More
Sino Biological Recombinant Human OX40 Ligand/TNFSF4 Protein (ECD, mFc Tag)
Supplier:  Sino Biological 13127H04H1
Encompass_Preferred
A DNA sequence encoding the human TNFSF4 (NP_003317.1) (Gln51-Leu183) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-255-4313