Learn More
Sino Biological Recombinant Human 4-1BB Ligand/TNFSF9 Protein (His Tag), Trimer, MALS-verified
Catalog No. 502993169
Encompass_Preferred
A DNA sequence encoding the Human TNFSF9 (NP_003802.1) (Arg71-Glu254) was expressed, with a polyhistidine tag at the N-terminus. This construct has a trimer design.
Catalog No. 50-299-3169 Supplier Sino Biological Supplier No. 15693H07H2100
A DNA sequence encoding the Human TNFSF9 (NP_003802.1) (Arg71-Glu254) was expressed, with a polyhistidine tag at the N-terminus. This construct has a trimer design.