Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TNIP3 Synthetic Peptide

Catalog No. PEP0912 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0912 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0912 Supplier Invitrogen™ Supplier No. PEP0912
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 17 amino acids near the carboxy terminus of human ABIN3. PEP-0912 can be used as a blocking peptide with polyclonal antibody PA5-20798.

The nuclear factor NF-kappa-B plays key roles in development and immunity. ABIN3 (A20-binding inhibitor of NF-kappa-B activation 3), also known as TNFAIP3-interacting protein 3 (TNIP3), is a novel negative feedback regulator of LPS-induced NF-kappa-B activation. ABIN3 is a 39 kDa protein that negatively regulates NF-kappa-B activation in response to TNF and LPS. ABIN3 is highly expressed in brain, thymus, lymph node, lung and fetal liver, with low expression in kidney, bone marrow. Through its interaction with A20, ABIN3 interferes with TRAF2-mediated transactivation signals and NF-kappa-B inhibition is mediated by the ABIN-homology domain 2. ABIN3 has been found to be induced by Listeria infection and can be slightly downregulated by dexamethasone. Enhanced expression of ABIN3 in monocytes is associated with sepsis. Thus, ABIN3 is an IL-10-induced gene product capable of attenuating NF-kappa-B in human macrophages yet is inoperative in mice and represents a basis for species-specific differences in IL-10 actions. At least four isoforms of ABIN3 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 79931
Species Human
Purification Method purified
Accession Number Q96KP6
Common Name TNIP3
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 9030611K07Rik; A20-binding inhibitor of NF-kappa-B activation 3; ABIN3; ABIN-3; ABIN-3 beta; LIND; Listeria induced; listeria-induced gene protein; TNFAIP3 interacting protein 3; TNFAIP3-interacting protein 3; TNFAIP3-interacting protein 3 beta; Tnip3; TNIP3 beta
Gene Symbol TNIP3
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.