Learn More
BIOHIPPO Human TNNC1 Microsample
SDP
Catalog No. 5000314516
Encompass
Human TNNC1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TNNC1 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 27 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TNNC1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TNNC1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TNNC1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14516 Supplier BIOHIPPO Supplier No. BHE15211405
Human TNNC1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TNNC1 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 27 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TNNC1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TNNC1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TNNC1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color