Learn More
Sino Biological Anti-Troponin C Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200106T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Troponin C / TNNC1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3977