Learn More
Sino Biological Anti-Troponin C Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11011T28100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Troponin C/TNNC1 (rh Troponin C/TNNC1; Catalog#11011-HNAE; NP_003271.1; Met1-Glu161). Troponin C/TNNC1 specific IgG was purified by Human Troponin C/TNNC1 affinity chromatography.
Catalog No. 50-255-4982