Learn More
BIOHIPPO Human TNNI1 Microsample
SDP
Catalog No. 5000314984
Encompass
Human TNNI1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TNNI1 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 46 88-3000 pg/mL with a sensitivity of 16 94 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TNNI1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TNNI1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TNNI1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14984 Supplier BIOHIPPO Supplier No. BHE15211722
Human TNNI1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TNNI1 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 46 88-3000 pg/mL with a sensitivity of 16 94 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TNNI1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TNNI1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TNNI1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color