Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TNPO3 Synthetic Peptide

Catalog No. PEP0671
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0671 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0671 Supplier Invitrogen™ Supplier No. PEP0671
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 18 amino acids near the amino terminus of human TNPO3. PEP-0671 can be used as a blocking peptide with polyclonal antibody PA5-20551.

TNPO3 is a member of the importin beta/transportin family of nuclear import receptors and is specific for the essential splicing factors termed SR proteins, binding specifically and directly to the arginine/serine (RS)-rich domains of these proteins. Further experiments demonstrated that this interaction occurs only when these RS domains are phosphorylated. More recently, TNPO3 was identified as an HIV dependency factor (HDF) and that depletion of TNPO3 resulted in HIV inhibition after reverse transcription but before integration, suggesting that TNPO3 may be an important drug target in HIV treatment. At least three isoforms of TNPO3 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 23534
Species Human
Purification Method purified
Accession Number Q9Y5L0
Common Name TNPO3
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 5730544L10Rik; C430013M08Rik; C81142; D6Ertd313e; Imp12; importin 12; Importin-12; IPO12; LGMD1F; limb girdle muscular dystrophy 1F (autosomal dominant); mKIAA4133; MTR10A; Tnpo3; transportin 3; transportin-3; Transportin-SR; TRNSR; TRN-SR; TRN-SR2
Gene Symbol TNPO3
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.