Learn More
Biomatik Corporation Human Toll Interleukin 1 Receptor Domain Containing Adaptor Protein (TIRAP) ELISA Kit
Supplier:  Biomatik Corporation EKU07743
Encompass_Preferred
Human Toll Interleukin 1 Receptor Domain Containing Adaptor Protein (TIRAP) ELISA Kit for detecting TIRAP in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.109ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P58753. Gene ID: 114609. Target Synonyms: Mal; Wyatt; MyD88 Adapter-Like; Adaptor protein Wyatt. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8727