Learn More
Biomatik Corporation Human Toll Like Receptor 9 (TLR9) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10963
Encompass_Preferred
Human Toll Like Receptor 9 (TLR9) High-sensitive ELISA Kit for detecting TLR9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NR96. Gene ID: 54106. Target Synonyms: CD289. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4720