Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TOM70 Synthetic Peptide

Catalog No. PEP0784 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0784 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0784 Supplier Invitrogen™ Supplier No. PEP0784
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 17 amino acids near the amino terminus of human TOM70. PEP-0784 can be used as a blocking peptide with polyclonal antibody PA5-20664.

Functional mitochondria require up to 1000 proteins to function properly, with 99% synthesized as precursors in the cytoplasm and transported into the mitochondria with the aid of cytosolic chaperones and mitochondrial translocators (import components). Proteins to be imported are chaperoned to the mitochondria by the cytosolic heat shock protein (cHSP70) and are immediately pursued by Translocators of the Outer Membrane (TOMs), followed by transient interactions of the unfolded proteins with Translocators of the Inner Membrane (TIMs). TOMM70A is ubiquitously expressed in human tissues and localizes in the mitochondria. TOMM70A could play a significant role in the import of nuclear-encoded mitochondrial proteins with internal targeting sites such as ADP/ATP carriers and the uncoupling proteins.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9868
Species Human
Purification Method purified
Accession Number O94826
Common Name TOM70
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 2610044B22Rik; D16Ium22; D16Ium22e; D16Wsu109e; FLJ90470; KIAA0719; Mitochondrial import receptor subunit TOM70; mitochondrial precursor proteins import receptor; mKIAA0719; Tom70; Tomm70; Tomm70a; Translocase of outer membrane 70 kDa subunit; translocase of outer mitochondrial membrane 70; translocase of outer mitochondrial membrane 70 homolog A; translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae); translocase of outer mitochondrial membrane 70 homolog A (yeast); translocase of outer mitochondrial membrane protein 70
Gene Symbol TOMM70
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.