Learn More
Amsbio LLC Human tonsil normal FFPE tissue block, 1 block
Supplier:  Amsbio LLC AMS6022/DEL
Encompass
amsbio supplies human tumor tissue samples from all major cancer types. Samples are collected under IRB procedures and consented to by donors. All tissues are processed by SOPs and reviewed by licensed pathologists. Stored at +4C but can be at room temp for a couple weeks. They’re shipped with gel packs.
Catalog No. NC1271017