Learn More
ACROBiosystems Human TOP2A / TOP2 Protein, Tag Free
Catalog No. 502055052
Encompass_Preferred
Recombinant Protein;100UG;Human TOP2A, Tag Free (TOA-H5117) is expressed from E.coli cells. It contains AA Ser 1354 - Ser 1473 (Accession # P11388-1).;This protein carries no "tag". The protein has a calculated MW of 12.8 kDa. The protein migrates as 20 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-205-5052 Supplier ACROBiosystems Supplier No. TOAH5117100UG
Recombinant Protein;100UG;Human TOP2A, Tag Free (TOA-H5117) is expressed from E.coli cells. It contains AA Ser 1354 - Ser 1473 (Accession # P11388-1).;This protein carries no "tag". The protein has a calculated MW of 12.8 kDa. The protein migrates as 20 kDa under reducing (R) condition (SDS-PAGE).