Learn More
TOPOGEN INC HUMAN TOPOISOMERASE II ASSAY
Catalog No. NC0646908
Encompass
Topo II Assay Kit. 100 assays. Storage: 4 C Short Term, -20 C Long Term
Catalog No. NC0646908 Supplier TOPOGEN INC Supplier No. TG10011
Topo II Assay Kit. 100 assays. Storage: 4 C Short Term, -20 C Long Term