Learn More
Biomatik Corporation Human Topoisomerase III (TOP3) ELISA Kit
Supplier:  Biomatik Corporation EKU07772
Encompass_Preferred
Human Topoisomerase III (TOP3) ELISA Kit for detecting TOP3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13472. Gene ID: 7156. Target Synonyms: TOP3A; DNA topoisomerase III alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6871